Anti-CSNK2B Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C2085-72P-APC
USBIC2085-72P-APCEA 884 EUR
USBIC2085-72P-APC
Anti-CSNK2B Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antilichamen
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: CSNK2B (casein kinase 2, beta polypeptide)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR