Anti-FLCN Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

F5730-01-FITC
USBIF5730-01-FITCEA 884 EUR
USBIF5730-01-FITC
Anti-FLCN Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antilichamen
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: FLCN (folliculin)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR