Anti-GALNS Goat polyclonal antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

G1024-01
USBIG1024-01EA 640 EUR
USBIG1024-01
Anti-GALNS Goat polyclonal antibody
Antilichamen
Recommended dilution: ELISA: 1: 64000; Western Blot: 0. 01-0. 03?g/ml

Type: Primary
Antigen: GALNS (galactosamine (N-acetyl)-6-sulfate sulfatase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR