Anti-Glucose Regulated Protein 94 Goat Polyclonal Antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

G3057-43A
USBIG3057-43AEA 640 EUR
USBIG3057-43A
Anti-Glucose Regulated Protein 94 Goat Polyclonal Antibody
Antilichamen
Recommended dilution: ELISA: 1:32000; Western Blot: 0. 01-0. 03?g/ml

Type: Primary
Antigen: Glucose Regulated Protein 94
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR