Anti-Glycerol-3-Phosphate Dehydrogenase Rabbit Polyclonal Antibody (Biotin)

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

G8154-72N-BIOT
USBIG8154-72N-BIOTEA 884 EUR
USBIG8154-72N-BIOT
Anti-Glycerol-3-Phosphate Dehydrogenase Rabbit Polyclonal Antibody (Biotin)
Antilichamen
Type: Primary
Antigen: Glycerol-3-Phosphate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Specificaties

antigeen symbool GPDH
Environmentally Preferable
Type antilichaam Primair,Glycerol-3-Phosphate Dehydrogenase
Type antilichaam Primair,Glycerol-3-Phosphate Dehydrogenase
Clonality Polyklonaal
Conjugatie Biotine
Reactivity Human,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitoop N-terminal
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR