Anti-IgG Chicken Polyclonal Antibody (Rhodamine)

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

I1904-45K7
USBII1904-45K7EA 258 EUR
USBII1904-45K7
Anti-IgG Chicken Polyclonal Antibody (Rhodamine)
Antilichamen
Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR