Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

I1904-53J5
USBII1904-53J5EA 369 EUR
USBII1904-53J5
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein))
Antilichamen
Recommended dilution: Immunochemistry: 1:20-1:5000

Type: Primary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Specificaties

antigeen symbool IgG
Environmentally Preferable
Type antilichaam Primair
Clonality Polyklonaal
Conjugatie FITC (Fluoresceïne-isothiocyanaat)
Reactivity Rabbit
Host Goat
Isotype
ImmunoChemie Yes
Flowcytometrie Yes
Formulier liquid
Opslagbuffer 10mMsodium phosphate, 0,15M sodium chloride, pH 7,2, 1% BSA, Protease/IgG free, 0,05% sodium azide.
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR