Anti-Ketamine Mouse Monoclonal Antibody [clone: 11C222]

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

K0542-01
USBIK0542-01EA 823 EUR
USBIK0542-01
Anti-Ketamine Mouse Monoclonal Antibody [clone: 11C222]
Antilichamen
Type: Primary
Antigen: Ketamine
Clonality: monoclonal
Clone: 11C222
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR