Anti-Luteinizing Hormone Rabbit Polyclonal Antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L7500-04O-100
USBIL7500-04O-100EA 692 EUR
USBIL7500-04O-100
Anti-Luteinizing Hormone Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: Luteinizing Hormone
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR