Anti-MHC Class 1 Chain-related Gene A Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M3886-75A-APC
USBIM3886-75A-APCEA 884 EUR
USBIM3886-75A-APC
Anti-MHC Class 1 Chain-related Gene A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antilichamen
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100; Flow Cytometry: 1:10-1:50; Immunofluorescence: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: MHC Class 1 Chain-related Gene A
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Specificaties

Environmentally Preferable
Type antilichaam Primair,MHC Class 1 Chain-related Gene A
Type antilichaam Primair,MHC Class 1 Chain-related Gene A
Clonality Polyklonaal
Conjugatie APC (Allophycocyanin)
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoFluorescentie Yes
Flowcytometrie Yes
Aminozuurgetal 75 to 105
Formulier liquid
Purification purified by Protein A chromatography
Opslagbuffer PBS, pH 7,2, contains ~0,09% sodium azide
Opslagtemperatuur Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR