Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody (Biotin)

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M9750-19B
USBIM9750-19BEA 605 EUR
USBIM9750-19B
Anti-Mycoplasma pneumoniae Rabbit Polyclonal Antibody (Biotin)
Antilichamen
Type: Primary
Antigen: Mycoplasma pneumoniae
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR