Anti-MYOD1 Rabbit Polyclonal Antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M9780-01C
USBIM9780-01CEA 379 EUR
USBIM9780-01C
Anti-MYOD1 Rabbit Polyclonal Antibody
Antilichamen
Recommended dilution: Western Blot: 1:500-1:1000

Type: Primary
Antigen: MYOD1 (myogenic differentiation 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR