Anti-POU2F1 Goat Polyclonal Antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P5250-01K
USBIP5250-01KEA 640 EUR
USBIP5250-01K
Anti-POU2F1 Goat Polyclonal Antibody
Antilichamen
Recommended dilution: ELISA: 1:4000; Western Blot: 0.5-2?g/ml

Type: Primary
Antigen: POU2F1 (POU class 2 homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR