Anti-PPARA Rabbit Polyclonal Antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P5505-07A
USBIP5505-07AEA 613 EUR
USBIP5505-07A
Anti-PPARA Rabbit Polyclonal Antibody
Antilichamen
Recommended dilution: ELISA: 1:8000-1:32000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: PPARA (peroxisome proliferator-activated receptor alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Specificaties

antigeen symbool PPARA
Environmentally Preferable
Type antilichaam Primair,peroxisome proliferator-activated receptor alpha
Type antilichaam Primair,peroxisome proliferator-activated receptor alpha
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitoop N-terminal
Formulier liquid
Purification purified by immunoaffinity chromatography
Concentratie ~1 mg/ml
Opslagbuffer PBS, pH 7,2, 0,09% sodium azide.
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR