Anti-Progesterone Mouse Monoclonal Antibody [clone: 10F313]

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P9006-01A
USBIP9006-01AEA 681 EUR
USBIP9006-01A
Anti-Progesterone Mouse Monoclonal Antibody [clone: 10F313]
Antilichamen
Recommended dilution: ELISA: 1:5000-1:15000

Type: Primary
Antigen: Progesterone
Clonality: monoclonal
Clone: 10F313
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR