Anti-Progesterone Mouse Monoclonal Antibody [clone: 10F318]

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P9006-01G
USBIP9006-01GEA 460 EUR
USBIP9006-01G
Anti-Progesterone Mouse Monoclonal Antibody [clone: 10F318]
Antilichamen
Type: Primary
Antigen: Progesterone
Clonality: monoclonal
Clone: 10F318
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Specificaties

antigeen symbool Progesterone
Environmentally Preferable
Type antilichaam Primair
Clonality Monoklonaal
Conjugatie Onconjugeerd
Clone 10F318
Reactivity
Host Mouse
Isotype IgG1
ELISA Yes
Formulier liquid
Purification purified by Protein A chromatography
Concentratie ~1 mg/ml
Opslagbuffer PBS, pH 7,2, 0,09% sodium azide.
Opslagtemperatuur Cool
Shipping temperature Ice
Tested applications Radio Immunoassay

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR