Anti-PTPN14 Chicken Polyclonal Antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P9109-30
USBIP9109-30EA 607 EUR
USBIP9109-30
Anti-PTPN14 Chicken Polyclonal Antibody
Antilichamen
Recommended dilution: Western Blot: 1:500; Immunochemistry: 1:200;

Type: Primary
Antigen: PTPN14 (protein tyrosine phosphatase, non-receptor type 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR