Anti-RB1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

R1660-05K2-PE
USBIR1660-05K2-PEEA 884 EUR
USBIR1660-05K2-PE
Anti-RB1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antilichamen
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: RB1 (retinoblastoma 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR