Anti-Salmonella sp. Rabbit Polyclonal Antibody (Biotin)

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

S0060-29B
USBIS0060-29BEA 459 EUR
USBIS0060-29B
Anti-Salmonella sp. Rabbit Polyclonal Antibody (Biotin)
Antilichamen
Recommended dilution: Immunofluorescence: 1:1000

Type: Primary
Antigen: Salmonella sp.
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR