Anti-Shigella sp. Rabbit Polyclonal Antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

S1013-15A
USBIS1013-15AEA 478 EUR
USBIS1013-15A
Anti-Shigella sp. Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: Shigella sp.
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR