Reusable earplugs, uvex whisper
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2111.201 2111.237
UVEX2111.201EA 119 EUR
UVEX2111.201 UVEX2111.237
Reusable earplugs, uvex whisper
Oordoppen Herbruikbare oordoppen
The earplugs with cord conveniently are at hand, whenever required. Manufactured from dirt repellent materials these ear plugs are suitable for use in dirty working environments.
  • With cord; can be worn comfortably around the neck
  • Hygienic storage in an individual carrying case
  • Dirt repellent surface can be cleaned using soap and water
  • Maintain the wearer's comfort and safety
Suitable for low noise areas.
SNR: 23 dB
Certificaten: EN 352-2:2002