Uvex Sportstyle Planet Glasses
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9193395 9193295
UVEX9193395EA 20 EUR
UVEX9193395 UVEX9193295
Uvex Sportstyle Planet Glasses
Brillen
Modern eye protection. Uvex sportstyle represents a new generation of innovative safety spectacles a clean, sporty design combining maximum functionality and protection, these lightweight safety glasses are a joy for everyone.
  • Sporty, lightweight safety spectacles with contoured lens for a great fit on narrow or wider faces
  • Uvex protecting planet - mass-balanced PC lens, frame and packaging partially made from recycled material
  • Robust polycarbonate lens with proven uvex supravision coating technology - permanently anti-fog and scratch-resistant lens
More Product Information