Chemical resistant gloves, uvex rubiflex S NB27S
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9893002 9893005 9893059 9893730
UVEX9893002EA 205 EUR
UVEX9893002 UVEX9893005 UVEX9893059 UVEX9893730
Chemical resistant gloves, uvex rubiflex S NB27S
Handschoenen
These chemical protection gloves feature a reinforced cotton interlock lining with a full NBR coating. This makes the gloves suitable for many applications in which chemical, heat or abrasion resistance is required.
  • Good resistance to chemicals, acids, alkalis, mineral oils and solvents
  • Good heat insulation with reinforced supporting material
  • Extremely high flexibility
  • Good tactile feel
  • Outstanding wearer comfort and ergonomic fit
Typical applications include: Chemical industry, sandblasting, automotive industry, metal working and machine and toolMore Product Information