Torches
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2010090400 9910056800 9910084700 2010069690 2010094800 2010104600 2010104700 2010104800 2010117400 2010117500 2010122400
VARI2010090400EA 687 EUR
VARI2010090400 VARI9910056800 VARI9910084700 VARI2010069690 VARI2010094800 VARI2010104600 VARI2010104700 VARI2010104800 VARI2010117400 VARI2010117500 VARI2010122400
Torches
Lampen Zaklampen
These torches light easily and stay lit, reducing downtime. With even and stable plasma to deliver reproducible results.