HPLC columns, Polaris™ ChromSep complete cartridge systems
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A2000100C030 A2000100C046 A2000150C046 A2000250C030 A2000250C046 A2000CG3 A2001100C046 A2001150C046 A2001250C046 A2001CG3
VARIA2000100C030EA 748 EUR
VARIA2000100C030 VARIA2000100C046 VARIA2000150C046 VARIA2000250C030 VARIA2000250C046 VARIA2000CG3 VARIA2001100C046 VARIA2001150C046 VARIA2001250C046 VARIA2001CG3
HPLC columns, Polaris™ ChromSep complete cartridge systems
Kolommen voor chromatografie
The ChromSep 316 stainless steel column-housing hardware is a durable one-time investment. The cartridge columns can be combined to match the column length with the separation you need and minimise your analysis time.
  • Easy, no-dead-volume assembly
  • Ergonomic format with modular flexibility