SPE plates, OMIX 96-well VersaPlate™
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A57103 A57103A A57109 A57109A A57111 A57111A
VARIA57103EA 0 EUR
VARIA57103 VARIA57103A VARIA57109 VARIA57109A VARIA57111 VARIA57111A
SPE plates, OMIX 96-well VersaPlate™
Microplaten Platen voor vastefase-extractie
OMIX automation-friendly 96-well monolithic SPE plates are specially designed to process small samples. They offer small extraction beds with almost no dead volume. Elution is achieved with microlitre solvent volumes, allowing direct injection and improving assay speed and sample throughput. OMIX tips are highly amenable to ADME/DMPK bioanalysis applications.
*Tubes need to be inserted into a VersaPlate™ base plate
Order Now
Specificaties
Environmentally Preferable