Filter kits, 96-well, Captiva
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A5960002SK A5960045SK A5967045K A5960045K A5960002K
VARIA5960002SKEA 1150 EUR
VARIA5960002SK VARIA5960045SK VARIA5967045K VARIA5960045K VARIA5960002K
Filter kits, 96-well, Captiva
Microplaten Filtreerplaten
Faster than centrifugation and easily automated, Captiva's dual depth filtration media provide complete removal of precipitated proteins and outstanding resistance to sample clogging. The plates are also excellent for sample storage. All Captiva components are ultra clean, and rigorously tested to ensure against non specific binding.
Leveringsinformatie: Each starter kit includes 5× Captiva filter plates, 10× Duo Seal 96 96-well plate seals, 5× Captiva 96-deep well 1 ml collection plates, and 5× Captiva collection plate pierceable covers. Each replacement filter kit includes 5× CaptivaMore Product Information