Particle traps for use with PLOT columns
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5181-3351 5181-3352 CP4788 CP4789 CP4795 CP4018 CP4017 CP4016
HEWL5181-3351EA 269 EUR
HEWL5181-3351 HEWL5181-3352 VARICP4788 VARICP4789 VARICP4795 VARICP4018 VARICP4017 VARICP4016
Particle traps for use with PLOT columns
Vallen voor vacuümpompen
Though highly stabilised, it is impossible to guarantee that no particles will dislodge from the column wall. When used in valve-switching applications, the use of a particle trap can prevent scarring of the rotors.