J&W PLOT PT GC columns
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CP7565PT CP7536PT CP7539PT CP7538PT CP7518PT CP7515PT CP7568PT CP7534PT 19095P-UO4PT
VARICP7565PTEA 1880 EUR
VARICP7565PT VARICP7536PT VARICP7539PT VARICP7538PT VARICP7518PT VARICP7515PT VARICP7568PT VARICP7534PT HEWL19095P-UO4PT
J&W PLOT PT GC columns
Kolommen voor chromatografie
PLOT PT columns have an integral particle trap at each end, to eliminate the bother of connecting separate traps, and enable GC/MS with leak-free operation and no anxiety.
  • Enhance analytical capability in the lab using PLOT analyses with MS detection and Capillary Flow Technology (CFT)
  • No more hassles with unions or downstream filters: Our integrated particle trapping technology is incorporated into the column as one continuous length of fused silica tubing
  • Higher throughput: Reproducible flow restriction and increased stability reduce the need for method/