Size exclusion chromatography columns, PL aquagel-OH™ preparative
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PL1220-6130 PL1249-1120 PL1249-6100 PL1249-6101 PL1249-6120 PL1249-6140 PL1249-6150 PL1249-6160
VARIPL1220-6130EA 5100 EUR
VARIPL1220-6130 VARIPL1249-1120 VARIPL1249-6100 VARIPL1249-6101 VARIPL1249-6120 VARIPL1249-6140 VARIPL1249-6150 VARIPL1249-6160
Size exclusion chromatography columns, PL aquagel-OH™ preparative
Kolommen voor chromatografie
These PL aquagel-OH preparative columns are used in fractionation of wide variety of water-soluble samples based on their size in solution.
  • High loading maximises sample throughput
  • Up to 10 times scale-up maximises yield
These columns and associated guard columns enable rapid and convenient scale-up from analytical separations. The columns are packed with the same robust macroporous particles as the analytical column range.