Pre-weighted calibration kits, EasiVial™
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PL2070-0201 PL2070-0202 PL2010-0200 PL2010-0202 PL2010-0203 PL2010-0300 PL2010-0301 PL2010-0400 PL2010-0401 PL2010-0402 PL2010-0403 PL2020-0200 PL2020-0201 PL2020-0202 PL2070-0200 PL2080-0200 PL2080-0202 PL2080-0203
VARIPL2070-0201EA 520 EUR
VARIPL2070-0201 VARIPL2070-0202 VARIPL2010-0200 VARIPL2010-0202 VARIPL2010-0203 VARIPL2010-0300 VARIPL2010-0301 VARIPL2010-0400 VARIPL2010-0401 VARIPL2010-0402 VARIPL2010-0403 VARIPL2020-0200 VARIPL2020-0201 VARIPL2020-0202 VARIPL2070-0200 VARIPL2080-0200 VARIPL2080-0202 VARIPL2080-0203
Pre-weighted calibration kits, EasiVial™
Standaarden Standaarden voor chromatografie
The quickest and most convenient method to deliver an accurate 12 point column calibration for organic and aqueous GPC/SEC column calibration.
  • Pre-prepared, time saving product for rapid and reliable GPC column calibration
  • Eliminates tedious weighing procedures to improve calibration accuracy
  • Reduces solvent dispensing to limit risks associated with handling solvents
  • For conventional and multi detector GPC to maximize applicability
The mass of each polymer in the vial is accurately known, so that upon addition of a fixed volume of eluent, the solution is prepared at a precise concentration. It is ideal for both conventional and multi detector GPC calibration. Simple to prepare and manually inject or transfer to autosampler vials or place directly into a compatible autosampler.