Button Batteries, Silver Coin
Leverancier: VARTA GERAETEBATTERIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
394101111 397101111 392101111 395101111 390101111 393101111 00391101111 00301101111
VART394101111EA 4.7 EUR
VART394101111 VART397101111 VART392101111 VART395101111 VART390101111 VART393101111 VART00391101111 VART00301101111
Button Batteries, Silver Coin
Batterijen
Different button cell types in low and high-drain versions.
  • Low self-discharge rate
  • Mercury-free
  • Guaranteed high level performance and extended storage time of up to three years
  • Chemical system: Primary silver oxide
The VARTA Silver Coin cells are well known as watch batteries. The wide variety of battery sizes delivering reliable, long-lasting energy and meeting highest quality and performance demands. Constant energy supply, withstands shock, vibration and acceleration, the cells are mercury-free and suitable for a broad scope of watches, toys,More Product Information
Order Now
Specificaties
Environmentally Preferable