Medical tubing, Tygon® ND 100-65
Leverancier: Saint Gobain Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ADF00001 ADF00002 ADF00003 ADF00006 ADF00007 ADF00009 ADF00011 ADF00012 ADF00013 ADF00017 ADF00018 ADF00022 ADF00023 ADF00027 ADF00028 ADF00037 ADF00046 ADF00053 ADF00062 ADF02012 ADF02017 ADF02037
VERNADF00001EA 53 EUR
VERNADF00001 VERNADF00002 VERNADF00003 VERNADF00006 VERNADF00007 VERNADF00009 VERNADF00011 VERNADF00012 VERNADF00013 VERNADF00017 VERNADF00018 VERNADF00022 VERNADF00023 VERNADF00027 VERNADF00028 VERNADF00037 VERNADF00046 VERNADF00053 VERNADF00062 VERNADF02012 VERNADF02017 VERNADF02037
Medical tubing, Tygon® ND 100-65
Slangen Transfer Tubing
Tygon®, crystal clear.
  • Crystal clear for easy visualisation of fluid flow
  • Ideal for contact with bodily fluids
  • Non-wetting surface permits complete drainage
  • Meets USP Class VI criteria
  • REACH compliant
  • Withstands EtO, gas and gamma sterilisation
Developed specifically for use in medical applications.
Temperature resistance: –42 to +74 °C
Hardness (Shore durometer A): 65
Density: 1,19