Boiling stones
Leverancier: Saint Gobain Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
D1069103
VERND1069103EA 48.1 EUR
VERND1069103
Boiling stones
Kookstenen
Ultra-pure, virgin PTFE boiling stones promote gentle, efficient boiling and help prevent overheating, even of highly volatile liquids. Ideal for continous extraction methods. Stones minimise 'bumping' during reflux and distillation.
  • Chemical resistant
  • Ideal for boiling organic liquids
  • Minimises superheating
The boiling stones are not affected by acids, hydrocarbons, ketones, esters, alcohols or alkalis. They are easily cleaned and non-contaminating. Shows no trace of face acidity or alkalinity. They will not scratch or abrade glassware and leave noMore Product Information