Cleanroom doekjes, EvoControl
Leverancier: FHP VILEDA SCS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
142870 142856 142857 142854 142855
VILE142870EA 2070 EUR
VILE142870 VILE142856 VILE142857 VILE142854 VILE142855
Cleanroom doekjes, EvoControl
Schoonmaakdoekjes Cleanroom doekjes Droge cleanroom doekjes
These microfibre cloth are manufactured from endless Evolon® microfibres measuring 0,15 dtex (10 times finer than standard polyester microfibres). The unique fabric combines very good textile and mechanical properties. With one single wiping movement, 99,999% of all bacteria are removed from the surface, even without the use of disinfectants.
  • Designed to hold the right amount of liquid, distributing it readily and evenly onto the surface
  • Microfibres reach even into microscopic surface pores and thus help remove contamination thouroughly
  • High contact area
More Product Information