Reagent reservoirs, disposable
Leverancier: VISTALAB TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3054-1003 3054-1005 3054-1004 4054-1004 3054-1001 3054-1002 3054-1006 3054-1007 3054-2000 3054-2001 3054-2002 3054-2003 3054-2006 3054-2007 3054-3001 4054-1000 4054-1002 4054-1006 3054-1013 3054-2013
VIST3054-1003EA 312 EUR
VIST3054-1003 VIST3054-1005 VIST3054-1004 VIST4054-1004 VIST3054-1001 VIST3054-1002 VIST3054-1006 VIST3054-1007 VIST3054-2000 VIST3054-2001 VIST3054-2002 VIST3054-2003 VIST3054-2006 VIST3054-2007 VIST3054-3001 VIST4054-1000 VIST4054-1002 VIST4054-1006 VIST3054-1013 VIST3054-2013
Reagent reservoirs, disposable
Reagensreservoirs
These reservoirs made of PS feature a trough-within-a-trough design to provide the lowest residual volume in the industry, meaning maximal reagent recovery.
  • Pour-off spouts at every corner
  • Wider base for stability
  • Free of animal products
  • Sterile reservoirs are certified RNase/DNase free and non pyrogenic