Safety wash bottles, wide neck, VITsafe™, with printed name
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1353859 1351859 1352859 1352879 1352889 1352969 1352979 1452989 1353819 1353839 1353849 1353869 1453829 1352909 1352939 1352949 1352899
VITL1353859EA 15.5 EUR
VITL1353859 VITL1351859 VITL1352859 VITL1352879 VITL1352889 VITL1352969 VITL1352979 VITL1452989 VITL1353819 VITL1353839 VITL1353849 VITL1353869 VITL1453829 VITL1352909 VITL1352939 VITL1352949 VITL1352899
Safety wash bottles, wide neck, VITsafe™, with printed name
Flessen Spuitflessen
Bottles made from LDPE (or PP for Acetone and MEK), spray tube from PP. With coloured VENT-CAP which supports easy visual identification of the media.
  • Material name in German, English, French and Spanish
  • Chemical formula, CAS No, hazard pictogram, signal word
  • Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
More safety due to the durable safety imprint in accordance to Directive (EC) No. 1272/2008 (GHS). Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube. PracticallyMore Product Information