Stands for Imhoff sedimentation cones
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
81056
VITL81056EA 147 EUR
VITL81056
Stands for Imhoff sedimentation cones
Staafstatieven
Sedimentation racks, made of PMMA. Holds two Imhoff sedimentation cones. Base plate with depression for exact vertical positioning of the sedimentation cone.
Order Now
Specificaties
Environmentally Preferable