VWR® flessen, smalle hals, met schroefdraad
Leverancier: LAB3 OWN PRODUCTION
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VWRLSTL12BEA 1670 EUR
VWRLSTL12B VWRLSTL18B VWRLSTL17B
VWR® flessen, smalle hals, met schroefdraad
Flessen Glazen flessen
Glass, clear or amber.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • Fitted White PTFE lined cap and label where stated
Excellent for sampling, storing and transporting liquids. Amber coloured to protect light-sensitive materials.