Cholesterol

Leverancier: Wako
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

413-43995
WAKO413-43995EA 0 EUR
WAKO413-43995
Cholesterol
Cholesterol
Formule: C₂₇H₄₆O
Kookpunt: 360 °C (1013 hPa)
Smeltpunt: 147…150 °C
Dichtheid: 1,07 g/cm³ (20 °C)
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00003646
CAS nummer: 57-88-5
EINECS: 200-353-2
Merck Index: 13,02221

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR