Sentry guard cartridges, XBridge™
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
186003064 186003061 186003060 186003079 186003082
WATE186003064EA 0 EUR
WATE186003064 WATE186003061 WATE186003060 WATE186003079 WATE186003082
Sentry guard cartridges, XBridge™
Kolommen voor chromatografie
XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1 to 12). The trifunctionally bonded C18 ligand provides with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed. The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent.