PC membranes, Whatman™ Cyclopore™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7060-4714 10417606 10417612 10417701 10417712 10418450 10418550 10419406 10419412
516-4556EA 184 EUR
516-4556 WHAT10417606 WHAT10417612 WHAT10417701 WHAT10417712 WHAT10418450 WHAT10418550 WHAT10419406 WHAT10419412
PC membranes, Whatman™ Cyclopore™
Filters Membraanfilters
Cytiva's Whatman Cyclopore polycarbonate membranes are true pore size microporous membranes featuring the sharp cut-off and reproducible microfiltration characteristics of track-etched membranes.
  • Low affinity for stains improves visibility under a microscope
  • Low nonspecific protein binding minimizes sample loss of proteinaceous samples
  • Narrow micropore size distribution and high pore density support rapid surface capture of analyte
  • Negligible absorption and adsorption of filtrate; nonhygroscopic
  • No particle shedding enables clean filtrate
  • Biologically
More Product Information