Membranes, AE, Whatman™

Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10549916
WHAT10549916EA 295 EUR
WHAT10549916
Membranes, AE, Whatman™
Filters Membraanfilters
These are unbacked nitrocellulose membranes for use in a lateral flow assay.

  • Unbacked, allowing either belt or air side of the membrane to be used
  • Enables assay suitability tests of both air and belt side of the membrane
  • Constructed of 100% nitrocellulose

These AE membranes offer a higher level of purity and performance compared to post-treated materials. The AE membranes have been used extensively since the development of the original lateral-flow tests.

Capillary flow rate: 160 - 210 s/4 cm
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR