Bottles, narrow neck, glass, safety coated, clear/amber, round, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
220975 220935 220936 220945 220946 220955 220925 220924 220976 220956 220735 220724 220725 220926 220736 220745 220746 220755 220756 220775 220776 220726
WHEA220975EA 503 EUR
WHEA220975 WHEA220935 WHEA220936 WHEA220945 WHEA220946 WHEA220955 WHEA220925 WHEA220924 WHEA220976 WHEA220956 WHEA220735 WHEA220724 WHEA220725 WHEA220926 WHEA220736 WHEA220745 WHEA220746 WHEA220755 WHEA220756 WHEA220775 WHEA220776 WHEA220726
Bottles, narrow neck, glass, safety coated, clear/amber, round, WHEATON®
Flessen Glazen flessen
Bottles, safety coated with PVC-based plastisol.Made from clear soda-lime glass that conforms to USP Type III requirements.
  • Non-slip coating allows for greater ease and stability when handling wet or dry
  • Autoclaving / sterilizing bottle is not recommended
  • Very good chemical resistance
  • Amber coloured for light sensitive substances
Plastisol coats exterior of bottle retaining contents if bottle is accidentally broken.
Certificaten: These bottles help meet OSHA requirements