Crimpers and decappers, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
224357 224307 224322 224323 224316
WHEA224357EA 1200 EUR
WHEA224357 WHEA224307 WHEA224322 WHEA224323 WHEA224316
Crimpers and decappers, WHEATON®
Capsuleer- en decapsuleertangen
These crimpers and decappers are used to attach and detach 30 mm aluminium seals.