Tubing, silicone or FKM, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
374311 374305 374313 374314 374316 374318 374319 374310 374306 374308
WHEA374311EA 237 EUR
WHEA374311 WHEA374305 WHEA374313 WHEA374314 WHEA374316 WHEA374318 WHEA374319 WHEA374310 WHEA374306 WHEA374308
Tubing, silicone or FKM, WHEATON®
Slangen Transfer Tubing
Silicone, translucent or FKM (Viton), black.
Silicone tubing
Very good temperature resistance
Autoclavable (121 °C)
Limited chemical resistance when used with concentrated acids
FKM (Viton) tubing
FKM tubing is known for its resistance to oils and chemicals, this remarkable material retains its good mechanical and chemical resistance properties at extremely high temperatures better than any other known elastomer. FKM is also a standout material in its resistance to weather-related aging and ozone.