Pasteur pipettes, LDPE, general purpose, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
711115 726129 711117S/10 711117S/20 711116 726128 711118S/10 711118S/20 711117F 711117F1S 711117F20S 711230 711117MF/3000
WHEA711115EA 288 EUR
WHEA711115 WHEA726129 WHEA711117S/10 WHEA711117S/20 WHEA711116 WHEA726128 WHEA711118S/10 WHEA711118S/20 WHEA711117F WHEA711117F1S WHEA711117F20S WHEA711230 WHEA711117MF/3000
Pasteur pipettes, LDPE, general purpose, WHEATON®
Pipetten Pasteur-/transferpipetten
These single use Pasteur pipettes are manufactured from non-toxic shatterproof LDPE.
  • Ideal for safe and easy transfer of liquid samples
  • Sterile and non-sterile versions in a range of packaging formats
  • Graduations on 1 and 3 ml pipettes for ease of volume control
Narrow stem pipettes can be used as a freeze vial when bulb is fully heat sealed. Jumbo pipettes are ideal for larger volume sample transfer and accessing liquids in taller vessels.