Bottles, wide neck, square, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
W216871 W216882 W216879 W216880 W216881 W216873 W216874 W216875 W216876 W216877 W216878 W216872 W216889 W216898
WHEAW216871EA 114 EUR
WHEAW216871 215-1680 215-1697 215-1698 215-1699 WHEAW216873 WHEAW216874 WHEAW216875 WHEAW216876 WHEAW216877 WHEAW216878 WHEAW216872 WHEAW216889 WHEAW216898
Bottles, wide neck, square, WHEATON®
Flessen Glazen flessen
Square shape glass bottles provides maximum storage space.
  • Ideal for solvent, chemical or sample storage
  • Clear glass helps for easy sample observation
  • Made of type III soda-lime glass