Erlenmeyer shake flasks, with DuoCap, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
WPFPC0125S WPFPC2000S WPFPC1000S WPFBC2000S WPFPC0500S WPFPC3000S WPFBC0250S WPFBC0500S WPFBC1000S
WHEAWPFPC0125SEA 549 EUR
WHEAWPFPC0125S WHEAWPFPC2000S WHEAWPFPC1000S WHEAWPFBC2000S WHEAWPFPC0500S WHEAWPFPC3000S WHEAWPFBC0250S WHEAWPFBC0500S WHEAWPFBC1000S
Erlenmeyer shake flasks, with DuoCap, WHEATON®
Kolven Erlenmeyers
These Erlenmeyer flasks are made of lexan polycarbonate resin that has outstanding mechanical, optical, electrical and thermal properties.
  • Manufactured in a class 10000 cleanroom
  • DuoCAP® allows the option of sterile air exchange or a leak resistant seal
  • Certified pyrogen-, DNase- and RNase-free
  • Gamma irradiated, sterile to SAL 10-6
  • Re-autoclavable and reusable
  • Supports both aerobic and anaerobic culture
The resin conforms to flame retardant standards, is scratch resistant and retains clarity after gamma irradiation. It also meets the requirements ofMore Product Information