Stopcock, three-way, acc. to Czako
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0.268.001
WITG0.268.001EA 62.1 EUR
WITG0.268.001
Stopcock, three-way, acc. to Czako
Afsluitkranen
Stopcocks made of borosilicate glass.
  • With capillary side arms
  • Solid glass key
  • Good chemical and temperature resistance
Certificaten: DIN 12563